6-Cyano-2-naphthol

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 6-Hydroxy-2-naphthonitrile6-Cyano-2-naphthol

ACTIAS31510G5EA 38.53 CHF
ACTIAS31510G10 ACTIAS31510G5 ACTIAS31510G1
6-Cyano-2-naphthol
6-Cyano-2-naphthol
Formula: C₁₁H₇NO
MW: 169.1794 g/mol
MDL Number: MFCD01863480
CAS Number: 52927-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR