4-Methylumbelliferone
Supplier: LGC Standards PROMOCHEM
Synonyms: β-MethylumbelliferoneHymecromone7-Hydroxy-4-methylcoumarin4-MU
PROOMM0294.00EA 2280 CHF
PROOMM0294.00
4-Methylumbelliferone
4-Methylumbelliferone
Formula: C₁₀H₈O₃
MW: 176.17 g/mol
MDL Number: MFCD00006866
CAS Number: 90-33-5
EINECS: 201-986-7