4-Chloro-6-methoxypyrimidine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS50161G5EA 0 CHF
ACTIAS50161G5 ACTIAS50161G25 ACTIAS50161G1
4-Chloro-6-methoxypyrimidine
4-Chloro-6-methoxypyrimidine
Formula: Câ‚…Hâ‚…ClNâ‚‚O
MW: 144.5590 g/mol
MDL Number: MFCD06200211
CAS Number: 26452-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR