2,4-Dichlorchinolin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS1439G5EA 0 CHF
ACTIAS1439G5 ACTIAS1439G1
2,4-Dichlorchinolin
2,4-Dichloroquinoline
Formel: C₉H₅Cl₂N
MW: 198,0500 g/mol
MDL Number: MFCD00023939
CAS-Nummer: 703-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR