2-Pyridylacetonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Pyridineacetonitrile

ACTIAS21238G25EA 0 CHF
ACTIAS21238G25 ACTIAS21238G100
2-Pyridylacetonitrile
2-Pyridylacetonitrile
Formula: C₇H₆N₂
MW: 118.14
MDL Number: MFCD00006346
CAS Number: 2739-97-1
EINECS: 220-364-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR