5-Bromo-2-(methylthio)pyrimidine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 5-Bromo-2-methylthiopyrimidine

ACTIAS50165G5EA 76.01 CHF
ACTIAS50165G25 ACTIAS50165G5 ACTIAS50165G1
5-Bromo-2-(methylthio)pyrimidine
5-Bromo-2-(methylthio)pyrimidine
Formula: Câ‚…Hâ‚…BrNâ‚‚S
MW: 205.0756 g/mol
MDL Number: MFCD00187875
CAS Number: 14001-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR