6-Methylpurine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS30797G1EA 0 CHF
ACTIAS30797G1 ACTIAS30797G5
6-Methylpurine
6-Methylpurine
Formula: C₆H₆N₄
MW: 134.1400 g/mol
MDL Number: MFCD00083663
CAS Number: 2004-03-7
EINECS: 217-903-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR