3-(Methoxycarbonyl)phenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-(Methoxycarbonyl)benzeneboronic acid

ACTIAS50860G5EA 0 CHF
ACTIAS50860G25 ACTIAS50860G1 ACTIAS50860G5
3-(Methoxycarbonyl)phenylboronic acid
3-(Methoxycarbonyl)phenylboronic acid
Formula: C₈H₉BO₄
MW: 179.97 g/mol
MDL Number: MFCD02093046
CAS Number: 99769-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR