(±)-1,2,3,4-Tetrahydro-1-naphthol

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1,2,3,4-Tetrahydro-1-naphtholac-alpha-Tetralol1,2,3,4-Tetrahydronaphthalen-1-ol

ACTIAS34430G5EA 0 CHF
ACTIAS34430G5 ACTIAS34430G10 ACTIAS34430G1
(±)-1,2,3,4-Tetrahydro-1-naphthol
(±)-1,2,3,4-Tetrahydro-1-naphthol
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00001739
CAS Number: 529-33-9
EINECS: 208-459-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR