2-Chloro-4-hydrazinopyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31013G1EA 187 CHF
ACTIAS31013G1
2-Chloro-4-hydrazinopyridine
2-Chloro-4-hydrazinopyridine
Formula: C₅H₆ClN₃
MW: 143.58 g/mol
MDL Number: MFCD08460247
CAS Number: 700811-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR