Sarcosine methyl ester hydrochloride

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: H-Sar-OMe·HCl

ACTIAS24056G100EA 0 CHF
ACTIAS24056G100
Sarcosine methyl ester hydrochloride
Sarcosine methyl ester hydrochloride
Formula: C₄H₉NO₂·HCl
MW: 139.58 g/mol
MDL Number: MFCD00038876
CAS Number: 13515-93-0
EINECS: 236-853-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR