7-Methoxy-2-tetralone
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
3,4-Dihydro-7-methoxynaphthalen-2(1H)-one
, 7-Methoxy-3,4-dihydronaphthalen-2(1H)-one
ACTIAS7022G5EA
0
CHF
ACTIAS7022G25
ACTIAS7022G1
ACTIAS7022G5
7-Methoxy-2-tetralone
7-Methoxy-2-tetralone
Formula:
C₁₁H₁₂O₂ MW: 176.22 g/mol |
MDL Number:
MFCD00001730 CAS Number: 4133-34-0 EINECS: 223-954-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...