6-Methoxy-1-tetralon
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS7070G5EA
0
CHF
ACTIAS7070G25
ACTIAS7070G5
ACTIAS7070G1
6-Methoxy-1-tetralon
6-Methoxy-1-tetralon
Formel:
C₁₁H₁₂O₂ MW: 176,22 g/mol |
MDL Number:
MFCD00001695 CAS-Nummer: 1078-19-9 EINECS: 214-078-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...