2-Amino-4-(trifluormethyl)pyrimidin
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS22205G25EA
0
CHF
ACTIAS22205G25
ACTIAS22205G5
2-Amino-4-(trifluormethyl)pyrimidin
2-Amino-4-(trifluoromethyl)pyrimidine
Formel:
C₅H₄F₃N₃ MW: 163,1 g/mol |
MDL Number:
MFCD00828624 CAS-Nummer: 16075-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...