1-Methyl-2-piperidinmethanol
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS40084G100EA
0
CHF
ACTIAS40084G100
1-Methyl-2-piperidinmethanol
1-Methyl-2-piperidinmethanol
Formel:
C₇H₁₅NO MW: 129,2 g/mol |
MDL Number:
MFCD00006494 CAS-Nummer: 20845-34-5 EINECS: 244-078-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...