Methylisobutyrylacetat
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS20180G100EA
58
CHF
ACTIAS20180G500
ACTIAS20180G100
Methylisobutyrylacetat
Methylisobutyrylacetat
Formel:
C₇H₁₂O₃ MW: 144,17 g/mol |
MDL Number:
MFCD00040499 CAS-Nummer: 42558-54-3 EINECS: 418-900-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...