3-Piperidinemethanol

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-(Hydroxymethyl)piperidine

ACTIAS20681G25EA 0 CHF
ACTIAS20681G100 ACTIAS20681G25
3-Piperidinemethanol
3-Piperidinemethanol
Formula: C₆H₁₃NO
MW: 115.18 g/mol
MDL Number: MFCD00005997
CAS Number: 4606-65-9
EINECS: 225-013-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR