1-Bromo-3-methoxypropane

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS20520G100EA 89 CHF
ACTIAS20520G100 ACTIAS20520G500 ACTIAS20520G25
1-Bromo-3-methoxypropane
1-Bromo-3-methoxypropane
Formula: C₄H₉BrO
MDL Number: MFCD02258473
CAS Number: 36865-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR