2,3-Dibromothiophene

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40102G10EA 0 CHF
ACTIAS40102G10
2,3-Dibromothiophene
2,3-Dibromothiophene
Formula: Câ‚„Hâ‚‚Brâ‚‚S
MW: 241.93 g/mol
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –17.5 °C
Density: 2.2 g/cm³ (20 °C)
MDL Number: MFCD00005418
CAS Number: 3140-93-0
EINECS: 221-542-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR