2-Isopropylpyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS32819G5EA 0 CHF
ACTIAS32819G5 ACTIAS32819G10 ACTIAS32819G1
2-Isopropylpyridine
2-Isopropylpyridine
Formula: C₈H₁₁N
MW: 121.18 g/mol
MDL Number: MFCD00047438
CAS Number: 644-98-4
EINECS: 211-426-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR