2-Chloro-3-picoline

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 2-Chloro 3-methylpyridine2-Chloro-3-methylpyridine2-Chloro-β-picoline

ACTIAS40202G100EA 0 CHF
ACTIAS40202G100
2-Chloro-3-picoline
2-Chloro-3-picoline
Formula: C₆H₆ClN
MW: 127.57 g/mol
MDL Number: MFCD00234177
CAS Number: 18368-76-8
EINECS: 242-242-1
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR