2-Ethoxypyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40232G100EA 0 CHF
ACTIAS40232G100
2-Ethoxypyridine
2-Ethoxypyridine
Formula: C₇H₉NO
MW: 123.15 g/mol
MDL Number: MFCD00023464
CAS Number: 14529-53-4
EINECS: 238-553-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR