2,3-Diaminopyridin

Lieferant: ACTIVATE SCIENTIFIC GMBH
Achtung

ACTIAS7044G100EA 0 CHF
ACTIAS7044G100 ACTIAS7044G25
2,3-Diaminopyridin
2,3-Diaminopyridin
Formel: C₅H₇N₃
MW: 109,13 g/mol
Siedepunkt: 148…150 °C (5 mmHg)
Melting Pt: 110…112 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006319
CAS-Nummer: 452-58-4
EINECS: 207-200-9
UN: 2671
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR