2,3,5,6-Tetrafluoropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

ACTIAS34429G1EA 46.55 CHF
ACTIAS34429G1 ACTIAS34429G5 ACTIAS34429G10
2,3,5,6-Tetrafluoropyridine
2,3,5,6-Tetrafluoropyridine
Formula: C₅HF₄N
MW: 151.06 g/mol
Boiling Pt: 98…99 °C (1013 hPa)
Density: 1.499 g/cm³ (20 °C)
MDL Number: MFCD00011734
CAS Number: 2875-18-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR