2-Chloro-3-methoxypyridine

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS21906G5EA 0 CHF
ACTIAS21906G5
2-Chloro-3-methoxypyridine
2-Chloro-3-methoxypyridine
Formula: C₆H₆ClNO
MW: 143.57 g/mol
MDL Number: MFCD03426022
CAS Number: 52605-96-6
EINECS: 258-039-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR