2-Chloro-3-nitropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS31487G100EA 0 CHF
ACTIAS31487G100
2-Chloro-3-nitropyridine
2-Chloro-3-nitropyridine
Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00006232
CAS Number: 5470-18-8
EINECS: 226-799-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR