2-Bromo-3-nitropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS1170G1EA 0 CHF
ACTIAS1170G1 ACTIAS1170G25 ACTIAS1170G5
2-Bromo-3-nitropyridine
2-Bromo-3-nitropyridine
Formula: C₅H₃BrN₂O₂
MW: 203 g/mol
MDL Number: MFCD00955613
CAS Number: 19755-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR