5-Bromo-2-iodopyridine

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS4013G100EA 0 CHF
ACTIAS4013KG1 ACTIAS4013G100
5-Bromo-2-iodopyridine
5-Bromo-2-iodopyridine
Formula: C₅H₃BrIN
MW: 283.89 g/mol
MDL Number: MFCD00691506
CAS Number: 223463-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR