3-Bromopyridine

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS51276G100EA 0 CHF
ACTIAS51276G500 ACTIAS51276G25 ACTIAS51276G100
3-Bromopyridine
3-Bromopyridine
Formula: C₅H₄BrN
MW: 158 g/mol
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: –27 °C
Density: 1.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006373
CAS Number: 626-55-1
EINECS: 210-952-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR