3,5-Dibromopyridine

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS50192G100EA 0 CHF
ACTIAS50192G100 ACTIAS50192G5 ACTIAS50192G25
3,5-Dibromopyridine
3,5-Dibromopyridine
Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 222 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00014634
CAS Number: 625-92-3
EINECS: 210-916-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR