5-Nitroisoquinoline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS20517G100EA 286 CHF
ACTIAS20517G100 ACTIAS20517G25
5-Nitroisoquinoline
5-Nitroisoquinoline
Formula: C₉H₆N₂O₂
MW: 174.16 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 111…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006905
CAS Number: 607-32-9
EINECS: 210-133-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR