4-Bromoisoquinoline

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS7036G100EA 0 CHF
ACTIAS7036G100 ACTIAS7036KG1
4-Bromoisoquinoline
4-Bromoisoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 42…42,5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006904
CAS Number: 1532-97-4
EINECS: 216-244-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR