3-Fluoro-2-nitropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS30499G5EA 108 CHF
ACTIAS30499G5 ACTIAS30499G1
3-Fluoro-2-nitropyridine
3-Fluoro-2-nitropyridine
Formula: C₅H₃FN₂O₂
MDL Number: MFCD04114127
CAS Number: 54231-35-5
EINECS: 207-260-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR