3-Brom-5-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS22146G5EA 0 CHF
ACTIAS22146G5
3-Brom-5-nitropyridin
3-Bromo-5-nitropyridine
Formel: C₅H₃BrN₂O₂
MDL Number: MFCD04114216
CAS-Nummer: 15862-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR