Methyl 2-methylnicotinate

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31638G5EA 0 CHF
ACTIAS31638G10 ACTIAS31638G5
Methyl 2-methylnicotinate
Methyl 2-methylnicotinate
Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00051961
CAS Number: 65719-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR