4-Iodopicolinic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Iodopyridine-2-carboxylic acid

ACTIAS51460G5EA 0 CHF
ACTIAS51460G5 ACTIAS51460G25 ACTIAS51460G1
4-Iodopicolinic acid
4-Iodopicolinic acid
Formula: C₆H₄INO₂
MW: 249.01 g/mol
MDL Number: MFCD03426666
CAS Number: 405939-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR