Methyl-6-aminonicotinat
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS7111G5EA
0
CHF
ACTIAS7111G100
ACTIAS7111G5
Methyl-6-aminonicotinat
Methyl-6-aminonicotinat
Formel:
C₇H₈N₂O₂ MW: 152,15 g/mol |
MDL Number:
MFCD00797844 CAS-Nummer: 36052-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...