Methyl-6-chloro-5-nitronicotinate

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS35404G5EA 178 CHF
ACTIAS35404G5 ACTIAS35404G1
Methyl-6-chloro-5-nitronicotinate
Methyl 6-chloro-5-nitronicotinate
Formula: C₇H₅ClN₂O₄
MW: 216.58 g/mol
MDL Number: MFCD01928341
CAS Number: 59237-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR