Ethyl-2-chloronicotinate

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Ethyl 2-chloropyridine-3-carboxylate

ACTIAS21733G100EA 0 CHF
ACTIAS21733G100 ACTIAS21733G25
Ethyl-2-chloronicotinate
Ethyl 2-chloronicotinate
Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
MDL Number: MFCD00674129
CAS Number: 1452-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR