5-Brom-2-hydroxy-3-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS40500G5EA 0 CHF
ACTIAS40500G5
5-Brom-2-hydroxy-3-nitropyridin
5-Brom-2-hydroxy-3-nitropyridin
Formel: C₅H₃BrN₂O₃
MW: 218,99 g/mol
MDL Number: MFCD00023473
CAS-Nummer: 15862-34-7
EINECS: 239-989-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR