Methyl-6-amino-5-bromonicotinate 98%

Supplier: Thermo Scientific
Warning

H33991.06EA 509 CHF
H33991.06
Methyl-6-amino-5-bromonicotinate 98%
Methyl 6-amino-5-bromonicotinate
Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD06659561
CAS Number: 180340-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR