Methyl-6-amino-5-bromonicotinate

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31072G1EA 0 CHF
ACTIAS31072G1 ACTIAS31072G5
Methyl-6-amino-5-bromonicotinate
Methyl 6-amino-5-bromonicotinate
Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
MDL Number: MFCD06659561
CAS Number: 180340-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR