2-Acetamido-5-fluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS21607G5EA 0 CHF
ACTIAS21607G5
2-Acetamido-5-fluorobenzoic acid
2-Acetamido-5-fluorobenzoic acid
Formula: C₉H₈FNO₃
MDL Number: MFCD00973900
CAS Number: 49579-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR