5-Bromopyridine-2-carbonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 5-Bromopicolinonitrile5-Bromo-2-cyanopyridine

ACTIAS4003G5EA 0 CHF
ACTIAS4003G25 ACTIAS4003G5
5-Bromopyridine-2-carbonitrile
5-Bromopyridine-2-carbonitrile
Formula: C₆H₃BrN₂
MW: 183.01 g/mol
MDL Number: MFCD00234144
CAS Number: 97483-77-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR