Bromobenzyl cyanide

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS36955G1EA 0 CHF
ACTIAS36955MG250 ACTIAS36955G1
Bromobenzyl cyanide
Bromobenzyl cyanide
Formula: C₈H₆BrN
MDL Number: MFCD01672925
CAS Number: 5798-79-8
EINECS: 227-348-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR