7-Iodoisatin

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS36770G1EA 0 CHF
ACTIAS36770MG250 ACTIAS36770G1
7-Iodoisatin
7-Iodoisatin
Formula: C₈H₄INO₂
MDL Number: MFCD01248407
CAS Number: 20780-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR