2,6-Dichloroisonicotinic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2,6-Dichloropyridine-4-carboxylic acid

ACTIAS1560G5EA 0 CHF
ACTIAS1560G5 ACTIAS1560G100 ACTIAS1560G25
2,6-Dichloroisonicotinic acid
2,6-Dichloroisonicotinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD00234147
CAS Number: 5398-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR