2,5-Dichloroisonicotinic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2,5-Dichloropyridine-4-carboxylic acid

ACTIAS1181G5EA 0 CHF
ACTIAS1181G5
2,5-Dichloroisonicotinic acid
2,5-Dichloroisonicotinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD00466640
CAS Number: 88912-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR