2,3-Dichloroisonicotinic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2,3-Dichloropyridine-4-carboxylic acid

ACTIAS25321G5EA 0 CHF
ACTIAS25321G5
2,3-Dichloroisonicotinic acid
2,3-Dichloroisonicotinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD01861977
CAS Number: 184416-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR