6-Chloroisatin

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31915G5EA 0 CHF
ACTIAS31915G5 ACTIAS31915G10
6-Chloroisatin
6-Chloroisatin
Formula: C₈H₄ClNO₂
MDL Number: MFCD00086347
CAS Number: 6341-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR